Assay Details
| Gene Symbol: LOC4344801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794248.2 | XP_015649734.1 | 16 - 17 | 3722 | 105 | |
| XM_015794249.2 | XP_015649735.1 | 15 - 16 | 3696 | 105 | ||
| GenBank mRNA | AK101719.1 | - | 15 - 16 | 3701 | 105 | |
More Information
Set Membership: |
|

LOC4344801
Genomic Map