Assay Details
| Gene Symbol: LOC4345269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795134.2 | XP_015650620.1 | 5 - 6 | 661 | 72 | |
| XM_015795135.2 | XP_015650621.1 | 5 - 6 | 661 | 72 | ||
| GenBank mRNA | AK064573.1 | - | 1 - 2 | 61 | 72 | |
More Information
Set Membership: |
|

LOC4345269
Genomic Map