Assay Details
| Gene Symbol: LOC4330517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768565.2 | XP_015624051.1 | 2 - 3 | 751 | 67 | |
| GenBank mRNA | AK060534.1 | - | 2 - 3 | 728 | 67 | |
| AK104296.1 | - | 2 - 3 | 728 | 67 | ||
More Information
Set Membership: |
|

LOC4330517
Genomic Map