Assay Details
| Gene Symbol: LOC4331292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776507.2 | XP_015631993.1 | 3 - 4 | 356 | 68 | |
| XM_015776508.2 | XP_015631994.1 | 3 - 4 | 349 | 68 | ||
| GenBank mRNA | AK072180.1 | - | 3 - 4 | 334 | 68 | |
| AK098927.1 | - | 3 - 4 | 270 | 68 | ||
More Information
Set Membership: |
|

LOC4331292
Genomic Map