Assay Details
| Gene Symbol: LOC4331292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776507.2 | XP_015631993.1 | 9 - 10 | 1016 | 81 | |
| XM_015776508.2 | XP_015631994.1 | 9 - 10 | 1009 | 81 | ||
| GenBank mRNA | AK072180.1 | - | 9 - 10 | 994 | 81 | |
| AK098927.1 | - | 9 - 10 | 930 | 81 | ||
More Information
Set Membership: |
|

LOC4331292
Genomic Map