Assay Details
| Gene Symbol: LOC4334930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779973.2 | XP_015635459.1 | 6 - 7 | 1177 | 77 | |
| GenBank mRNA | AK065187.1 | - | 5 - 6 | 1885 | 77 | |
| BT129204.1 | - | 2 - 3 | 222 | 77 | ||
More Information
Set Membership: |
|

LOC4334930
Genomic Map