Assay Details
| Gene Symbol: LOC4332216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777426.2 | XP_015632912.1 | 11 - 12 | 1773 | 90 | |
| XM_015777427.2 | XP_015632913.1 | 11 - 12 | 1729 | 90 | ||
| XM_015777428.2 | XP_015632914.1 | 11 - 12 | 1830 | 90 | ||
More Information
Set Membership: |
|

LOC4332216
Genomic Map