Assay Details
| Gene Symbol: LOC4333331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777567.2 | XP_015633053.1 | 2 - 3 | 702 | 79 | |
| GenBank mRNA | AK058432.1 | - | 2 - 3 | 393 | 79 | |
| AK099288.1 | - | 2 - 3 | 263 | 79 | ||
More Information
Set Membership: |
|

LOC4333331
Genomic Map