Assay Details
| Gene Symbol: LOC4342941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789861.2 | XP_015645347.1 | 1 | 642 | 87 | |
| GenBank mRNA | AK105204.1 | - | 1 | 648 | 87 | |
More Information
Set Membership: |
|

LOC4342941
Genomic Map