Assay Details
| Gene Symbol: LOC4346300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792811.2 | XP_015648297.1 | 4 - 5 | 76 | ||
| GenBank mRNA | AK061111.1 | - | 4 - 5 | 76 | ||
| AY345242.1 | - | 4 - 5 | 76 | |||
| AY569613.1 | - | 3 - 4 | 76 | |||
| AY581257.1 | - | 4 - 5 | 423 | 76 | ||
More Information
Set Membership: |
|

LOC4346300
Genomic Map