Assay Details
| Gene Symbol: EGLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194425.2 | NP_001181354.1 | 1017 | 75 | ||
| GenBank mRNA | JU332748.1 | - | 1004 | 75 | ||
| JV570013.1 | - | 995 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EGLN3
Genomic Map