Assay Details
| Gene Symbol: ECD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261188.1 | NP_001248117.1 | 1458 | 133 | ||
| XM_015147349.2 | XP_015002835.1 | 1480 | 133 | |||
| XM_015147350.2 | XP_015002836.1 | 1351 | 133 | |||
| XM_015147351.1 | XP_015002837.1 | 1357 | 133 | |||
| XM_028853440.1 | XP_028709273.1 | 1862 | 133 | |||
| XM_028853441.1 | XP_028709274.1 | 1837 | 133 | |||
| GenBank mRNA | JU323251.1 | - | 1459 | 133 | ||
| JU472438.1 | - | 1458 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ECD
Genomic Map