Assay Details
| Gene Symbol: SPPL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266692.1 | NP_001253621.1 | 1127 | 64 | ||
| GenBank mRNA | JU320525.1 | - | 1231 | 64 | ||
| JU472793.1 | - | 1178 | 64 | |||
| JV046164.1 | - | 1127 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPPL3
Genomic Map