Assay Details
| Gene Symbol: LSM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260685.1 | NP_001247614.1 | 377 | 140 | ||
| XM_015135932.2 | XP_014991418.1 | 397 | 140 | |||
| GenBank mRNA | JU320755.1 | - | 379 | 140 | ||
| JU472045.1 | - | 376 | 140 | |||
| JV045316.1 | - | 377 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LSM2
Genomic Map