Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124927.2 | XP_014980413.1 | 1327 | 74 | ||
| XM_015124928.2 | XP_014980414.1 | 1324 | 74 | |||
| XM_015124930.2 | XP_014980416.2 | 2044 | 74 | |||
| XM_028838851.1 | XP_028694684.1 | 1321 | 74 | |||
| GenBank mRNA | JU336100.1 | - | 1181 | 74 | ||
| JV591945.1 | - | 1218 | 74 | |||
| JV694552.1 | - | 1219 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SYT5
Genomic Map