Assay Details
| Gene Symbol: ADH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260627.1 | NP_001247556.1 | 72 | 155 | ||
| GenBank mRNA | JU473175.1 | - | 72 | 155 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260627.1 transcripts.
Set Membership: |
|

ADH1B
Genomic Map