Assay Details
Gene Symbol: LOC114674356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840866.1 | XP_028696699.1 | 1331 | 124 | ||
XM_028840867.1 | XP_028696700.1 | 998 | 124 | |||
XM_028840868.1 | XP_028696701.1 | 979 | 124 | |||
XM_028840870.1 | XP_028696703.1 | 807 | 124 |
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840864.1 | XP_028696697.1 | 244 | 124 | ||
GenBank mRNA | JU340450.1 | - | 98 | 124 | ||
JU470587.1 | - | 176 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840864.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
