Assay Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260588.1 | NP_001247517.1 | 283 | 110 | ||
| XM_015148591.2 | XP_015004077.2 | 357 | 110 | |||
| XM_028844607.1 | XP_028700440.1 | 231 | 110 | |||
| XM_028844611.1 | XP_028700444.1 | 208 | 110 | |||
| GenBank mRNA | JU335550.1 | - | 254 | 110 | ||
| JU476479.1 | - | 322 | 110 | |||
| JV044733.1 | - | 283 | 110 | |||
| JV551586.1 | - | 239 | 110 | |||
| JV690430.1 | - | 353 | 110 | |||
More Information
Set Membership: |
|

CTNNBIP1
Genomic Map