Assay Details
| Gene Symbol: GPRC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151162.2 | XP_015006648.1 | 1805 | 130 | ||
| XM_015151166.2 | XP_015006652.1 | 1955 | 130 | |||
More Information
Set Membership: |
|

GPRC5A
Genomic Map