Assay Details
| Gene Symbol: RBM46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139376.2 | XP_014994862.1 | 1713 | 113 | ||
| XM_015139377.1 | XP_014994863.1 | 1601 | 113 | |||
| XM_015139378.2 | XP_014994864.1 | 1527 | 113 | |||
| XM_015139380.2 | XP_014994866.1 | 2873 | 113 | |||
| XM_015139381.2 | XP_014994867.1 | 2813 | 113 | |||
More Information
Set Membership: |
|

RBM46
Genomic Map