Assay Details
| Gene Symbol: DCAF12L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193853.1 | NP_001180782.1 | 430 | 144 | ||
| XM_015128157.2 | XP_014983643.1 | 692 | 144 | |||
| XM_028841736.1 | XP_028697569.1 | 693 | 144 | |||
More Information
Set Membership: |
|

DCAF12L2
Genomic Map