Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113156.4 | XP_001113156.2 | 776 | 101 | ||
| GenBank mRNA | JU475137.1 | - | 661 | 101 | ||
| JV046733.1 | - | 651 | 101 | |||
More Information
Set Membership: |
|

MYCBP
Genomic Map