Assay Details
| Gene Symbol: PSENEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194826.1 | NP_001181755.1 | 211 | 87 | ||
| XM_002801186.3 | XP_002801232.1 | 283 | 87 | |||
| XM_015123558.2 | XP_014979044.1 | 283 | 87 | |||
| XM_028838309.1 | XP_028694142.1 | 249 | 87 | |||
| XM_028838310.1 | XP_028694143.1 | 269 | 87 | |||
| XM_028838311.1 | XP_028694144.1 | 231 | 87 | |||
| XM_028838312.1 | XP_028694145.1 | 200 | 87 | |||
| XM_028838313.1 | XP_028694146.1 | 217 | 87 | |||
| GenBank mRNA | JU320462.1 | - | 261 | 87 | ||
| JU473821.1 | - | 266 | 87 | |||
| JV045648.1 | - | 230 | 87 | |||

PSENEN
Genomic Map