Assay Details
| Gene Symbol: GMCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799303.3 | XP_002799349.3 | 515 | 110 | ||
| XM_028832419.1 | XP_028688252.1 | 540 | 110 | |||
| GenBank mRNA | JU321420.1 | - | 469 | 110 | ||
| JU472837.1 | - | 472 | 110 | |||
More Information
Set Membership: |
|

GMCL1
Genomic Map