Assay Details
| Gene Symbol: GPX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098032.4 | XP_001098032.2 | 531 | 93 | ||
| XM_028849439.1 | XP_028705272.1 | 502 | 93 | |||
| GenBank mRNA | JV652621.1 | - | 487 | 93 | ||
| JV652622.1 | - | 487 | 93 | |||
More Information
Set Membership: |
|

GPX8
Genomic Map