Assay Details
| Gene Symbol: PIGW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257700.1 | NP_001244629.1 | 46 | 84 | ||
| XM_028835742.1 | XP_028691575.1 | 382 | 84 | |||
| GenBank mRNA | JU323277.1 | - | 46 | 84 | ||
| JV058476.1 | - | 46 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257700.1, XM_028835742.1 transcripts.
Set Membership: |
|

PIGW
Genomic Map