Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134378.2 | XP_014989864.2 | 644 | 70 | ||
| XM_028845559.1 | XP_028701392.1 | 644 | 70 | |||
| GenBank mRNA | JU336662.1 | - | 614 | 70 | ||
| JU477462.1 | - | 629 | 70 | |||
| JV048418.1 | - | 626 | 70 | |||
| JV663184.1 | - | 622 | 70 | |||
More Information
Set Membership: |
|

ZNF394
Genomic Map