Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830632.1 | XP_028686465.1 | 104 | 118 | ||
| XM_028830634.1 | XP_028686467.1 | 104 | 118 | |||
| GenBank mRNA | JV522407.1 | - | 68 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028830632.1, XM_028830634.1 transcripts.
Set Membership: |
|

GPR61
Genomic Map