Assay Details
| Gene Symbol: LOC100423704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122127.2 | XP_014977613.2 | 33 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100423704| Gene Symbol: LOC100423704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122127.2 | XP_014977613.2 | 33 | 103 | ||
Set Membership: |
|