Assay Details
| Gene Symbol: MAGEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265825.1 | NP_001252754.1 | 209 | 100 | ||
| XM_028841693.1 | XP_028697526.1 | 291 | 100 | |||
| GenBank mRNA | JV636209.1 | - | 209 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265825.1, XM_028841693.1 transcripts.
Set Membership: |
|

MAGEC2
Genomic Map