Assay Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130750.1 | NP_570106.1 | 10 - 11 | 1076 | 136 | |
| GenBank mRNA | AF461260.1 | - | 10 - 11 | 1076 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P395642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44288 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P395642 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P395642 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P395642 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P395642 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91958 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34905 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38321 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86778 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10861161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861167 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861170 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861169 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861168 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861164 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10861157 | - | - |
| Rat Genome 230 2.0 Array | 1369486_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369486_at | - | - |
| Affymetrix HT Rat Focus Array | 1369486_at | - | - |
| Affymetrix HT Rat Focus Array | 1369486_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400017796.rn.1 | TC0400002514.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369486_PM_at | - | - |
| Rat Expression Set 230 | 1369486_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861157 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21170004 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373884 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64588 | - | - |

Asz1
Genomic Map