Assay Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138877.1 | NP_620232.1 | 1 - 2 | 58 | 89 | |
| GenBank mRNA | BC062066.1 | - | 1 - 2 | 74 | 5599255 | 89 |
| D00636.1 | - | 1 - 2 | 58 | 89 | ||
| J03867.1 | - | 1 - 2 | 53 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062066.1, D00636.1, J03867.1, NM_138877.1 transcripts.
Set Membership: |
|

Cyb5r3
Genomic Map