Assay Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031082.1 | NP_112344.1 | 4 - 5 | 491 | 64 | |
| XM_006254691.2 | XP_006254753.1 | 4 - 5 | 1731 | 64 | ||
| XM_008772177.1 | XP_008770399.1 | 5 - 6 | 493 | 64 | ||
| GenBank mRNA | L24116.1 | - | 4 - 5 | 491 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pggt1b
Genomic Map