Assay Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053583.2 | NP_446035.2 | 342 | 121 | ||
| GenBank mRNA | U92564.1 | - | 340 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the U92564.1 transcripts.
Set Membership: |
|

Zfp423
Genomic Map