Assay Details
| Gene Symbol: Pdzk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130401.1 | NP_569085.1 | 1 - 2 | 165 | 67 | |
| XM_006238620.2 | XP_006238682.1 | 2 - 3 | 186 | 67 | ||
| XM_008763968.1 | XP_008762190.1 | 2 - 3 | 358 | 67 | ||
| XM_008763969.1 | XP_008762191.1 | 2 - 3 | 186 | 67 | ||
| XM_008763970.1 | XP_008762192.1 | 2 - 3 | 171 | 67 | ||
| GenBank mRNA | AF402772.1 | - | 1 - 2 | 165 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdzk1ip1
Genomic Map