Assay Details
| Gene Symbol: Clmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173154.1 | NP_775177.1 | 6 - 7 | 912 | 114 | |
| GenBank mRNA | AF302047.1 | - | 6 - 7 | 912 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P102671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78006 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P102671 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P102671 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P102671 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P292495 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P102671 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31841 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81307 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65378 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78316 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53343 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 651 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66699 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10909345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909339 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909341 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10909338 | - | - |
| Rat Genome 230 2.0 Array | 1377639_at | - | - |
| Rat Genome 230 2.0 Array | 1376198_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377639_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376198_at | - | - |
| Affymetrix HT Rat Focus Array | 1377639_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376198_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377639_at | - | - |
| Affymetrix HT Rat Focus Array | 1376198_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800003145.rn.1 | TC0800000505.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376198_at | - | - |
| Rat Expression Set 230 | 1376198_PM_at | - | - |
| Rat Expression Set 230 | 1377639_at | - | - |
| Rat Expression Set 230 | 1377639_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10909338 | - | - |
| Rat Genome U34 Set | rc_AI008313_at | - | - |
| Rat Genome U34 Set | rc_AI011326_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21938248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365205 | - | - |
| RatRef-12 Expression BeadChip | ILMN_50162 | - | - |

Clmp
Genomic Map