Assay Details
| Gene Symbol: Kif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057200.2 | NP_476548.1 | 26 - 27 | 3028 | 61 | |
| XM_008764204.1 | XP_008762426.1 | 26 - 27 | 3034 | 61 | ||
| XM_008764205.1 | XP_008762427.1 | 24 - 25 | 2738 | 61 | ||
| GenBank mRNA | AB070355.1 | - | 26 - 27 | 3028 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif1b
Genomic Map