Assay Details
| Gene Symbol: Uts2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198133.2 | NP_937766.1 | 2 - 3 | 263 | 110 | |
| GenBank mRNA | AB116019.1 | - | 2 - 3 | 225 | 110 | |
| AY321314.1 | - | 2 - 3 | 263 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uts2b
Genomic Map