Assay Details
| Gene Symbol: Pcdhb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173099.1 | NP_775122.1 | 1 | 2854 | 65 | |
| XM_006254626.2 | XP_006254688.1 | 2 | 3441 | 65 | ||
| GenBank mRNA | L43592.1 | - | 1 | 2854 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcdhb12
Genomic Map