Assay Details
| Gene Symbol: Arhgap19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749116.3 | XP_003749164.1 | 3 - 4 | 431 | 90 | |
| XM_008760475.1 | XP_008758697.1 | 2 - 3 | 165 | 90 | ||
More Information
Set Membership: |
|

Arhgap19
Genomic Map