Assay Details
| Gene Symbol: Rsph10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013867.1 | NP_001013889.1 | 3 - 4 | 488 | 73 | |
| XM_006248861.2 | XP_006248923.1 | 3 - 4 | 1224 | 73 | ||
| GenBank mRNA | BC081935.1 | - | 3 - 4 | 488 | 7114952 | 73 |
| FQ212099.1 | - | 3 - 4 | 595 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsph10b
Genomic Map