Assay Details
| Gene Symbol: Msn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030863.1 | NP_110490.1 | 10 - 11 | 1346 | 58 | |
| GenBank mRNA | AF004811.1 | - | 10 - 11 | 1346 | 58 | |
| FQ230914.1 | - | 1433 | 58 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Msn
Genomic Map