Assay Details
| Gene Symbol: LOC100362572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767467.1 | XP_008765689.1 | 2 - 3 | 465 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100362572| Gene Symbol: LOC100362572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767467.1 | XP_008765689.1 | 2 - 3 | 465 | 74 | |
Set Membership: |
|