Assay Details
| Gene Symbol: LOC100911860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767354.1 | XP_008765576.1 | 7 - 8 | 1997 | 132 | |
| Gene Symbol: Usp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134885.1 | NP_001128357.1 | 8 - 9 | 1075 | 132 | |
| XM_008767258.1 | XP_008765480.1 | 8 - 9 | 1641 | 132 | ||
| XM_008767259.1 | XP_008765481.1 | 8 - 9 | 1536 | 132 | ||
| XM_008767260.1 | XP_008765482.1 | 8 - 9 | 1531 | 132 | ||
| XM_008767261.1 | XP_008765483.1 | 7 - 8 | 1997 | 132 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911860
Genomic Map