Assay Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772820.1 | XP_008771042.1 | 5 - 6 | 631 | 72 | |
| XM_008772821.1 | XP_008771043.1 | 5 - 6 | 625 | 72 | ||
More Information
Set Membership: |
|

Zfp523
Genomic Map