Assay Details
| Gene Symbol: Obp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128139.2 | NP_001121611.1 | 5 - 6 | 509 | 78 | |
| GenBank mRNA | DQ537493.1 | - | 5 - 6 | 511 | 78 | |
More Information
Set Membership: |
|

Obp2a
Genomic Map