Assay Details
| Gene Symbol: RGD1562029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767174.1 | XP_008765396.1 | 11 - 12 | 2325 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562029| Gene Symbol: RGD1562029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767174.1 | XP_008765396.1 | 11 - 12 | 2325 | 85 | |
Set Membership: |
|