Assay Details
| Gene Symbol: Ggta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145674.1 | NP_663707.1 | 3 - 4 | 333 | 98 | |
| XM_006234033.2 | XP_006234095.1 | 3 - 4 | 468 | 98 | ||
| XM_006234037.2 | XP_006234099.1 | 3 - 4 | 783 | 98 | ||
| XM_008761700.1 | XP_008759922.1 | 3 - 4 | 468 | 98 | ||
| XM_008761701.1 | XP_008759923.1 | 2 - 3 | 292 | 98 | ||
| GenBank mRNA | AF520589.1 | - | 3 - 4 | 333 | 98 | |
| BC166599.1 | - | 3 - 4 | 468 | 7932138 | 98 | |
| FQ222249.1 | - | 3 - 4 | 470 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ggta1
Genomic Map