Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767061.1 | XP_008765283.1 | 4 - 5 | 647 | 75 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA catabolic process, endonucleolytic
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, DNA incision, 3'-to lesion
biological_process
metabolic process
determination of adult lifespan
response to UV
UV protection
response to UV-C
multicellular organism growth
negative regulation of apoptotic process
nucleic acid phosphodiester bond hydrolysis
bubble DNA binding
molecular_function
DNA binding
double-stranded DNA binding
single-stranded DNA binding
catalytic activity
endonuclease activity
endodeoxyribonuclease activity
protein homodimerization activity
protein N-terminus binding
DNA catabolic process, endonucleolytic
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, DNA incision, 3'-to lesion
biological_process
metabolic process
determination of adult lifespan
response to UV
UV protection
response to UV-C
multicellular organism growth
negative regulation of apoptotic process
nucleic acid phosphodiester bond hydrolysis
bubble DNA binding
molecular_function
DNA binding
double-stranded DNA binding
single-stranded DNA binding
catalytic activity
endonuclease activity
endodeoxyribonuclease activity
protein homodimerization activity
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1019654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150905 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1019654 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1019654 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1019654 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P424587 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1019654 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31051 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10923015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923027 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923014 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923030 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923028 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10923012 | - | - |
| Rat Genome 230 2.0 Array | 1383953_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383953_at | - | - |
| Rat Genome 230 2.0 Array | 1398057_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398057_at | - | - |
| Affymetrix HT Rat Focus Array | 1383953_at | - | - |
| Affymetrix HT Rat Focus Array | 1383953_PM_at | - | - |
| Rat Expression Set 230 | 1383953_at | - | - |
| Rat Expression Set 230 | 1383953_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22386502 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355376 | - | - |

Ercc5
Genomic Map