Assay Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105883.1 | NP_001099353.1 | 8 - 9 | 1370 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110531 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029102 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029102 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029102 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P145290 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P270395 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35008 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49065 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98911 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10750990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750984 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10750982 | - | - |
| Rat Genome 230 2.0 Array | 1385316_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377714_at | - | - |
| Rat Genome 230 2.0 Array | 1380805_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377714_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397363_at | - | - |
| Rat Genome 230 2.0 Array | 1380805_at | - | - |
| Rat Genome 230 2.0 Array | 1385316_at | - | - |
| Rat Genome 230 2.0 Array | 1378027_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397363_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378027_at | - | - |
| Affymetrix HT Rat Focus Array | 1397363_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385316_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385316_at | - | - |
| Affymetrix HT Rat Focus Array | 1378027_at | - | - |
| Affymetrix HT Rat Focus Array | 1397363_at | - | - |
| Affymetrix HT Rat Focus Array | 1378027_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100009402.rn.1 | TC1100001543.rn.1 | RTA TC |
| Rat Expression Set 230 | 1377714_PM_at | - | - |
| Rat Expression Set 230 | 1385316_at | - | - |
| Rat Expression Set 230 | 1385316_PM_at | - | - |
| Rat Expression Set 230 | 1397363_at | - | - |
| Rat Expression Set 230 | 1378027_at | - | - |
| Rat Expression Set 230 | 1380805_at | - | - |
| Rat Expression Set 230 | 1378027_PM_at | - | - |
| Rat Expression Set 230 | 1377714_at | - | - |
| Rat Expression Set 230 | 1397363_PM_at | - | - |
| Rat Expression Set 230 | 1380805_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750982 | - | - |
| Rat Genome U34 Set | rc_AA925450_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21121258 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352327 | - | - |

Pvrl3
Genomic Map